Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPEF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024867.3 | NP_079143.3 | 26 | 3815 | ||
XM_005248376.3 | XP_005248433.1 | 26 | 3920 | |||
XM_005248377.3 | XP_005248434.1 | 26 | 3905 | |||
XM_011514135.2 | XP_011512437.1 | 26 | 3925 | |||
XM_011514136.2 | XP_011512438.1 | 26 | 3924 | |||
XM_011514137.2 | XP_011512439.1 | 26 | 3909 | |||
XM_011514138.2 | XP_011512440.1 | 26 | 3901 | |||
XM_011514139.2 | XP_011512441.1 | 25 | 3849 | |||
XM_011514140.2 | XP_011512442.1 | 25 | 3659 | |||
XM_011514141.2 | XP_011512443.1 | 27 | 4123 | |||
XM_011514142.1 | XP_011512444.1 | 26 | 3896 | |||
XM_017009880.1 | XP_016865369.1 | 25 | 3741 | |||
XM_017009881.1 | XP_016865370.1 | 26 | 4779 | |||
XM_017009882.1 | XP_016865371.1 | 25 | 3761 | |||
GenBank mRNA | AB051557.1 | - | 16 | 2156 | ||
AK027230.1 | - | 3 | 261 | |||
AK225586.1 | - | 3 | 261 | |||
AK225607.1 | - | 3 | 261 |