Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024899.3 | NP_079175.2 | 4 | 554 | ||
NR_073537.1 | 4 | 549 | ||||
XM_005258149.4 | XP_005258206.1 | 4 | 533 | |||
XM_017025981.1 | XP_016881470.1 | 4 | 533 | |||
XM_017025982.1 | XP_016881471.1 | 4 | 614 | |||
XM_017025983.1 | XP_016881472.1 | 4 | 533 | |||
XM_017025986.1 | XP_016881475.1 | 3 | 291 | |||
GenBank mRNA | AK022604.1 | - | 4 | 416 | ||
AK301964.1 | - | 3 | 291 | |||
BC026307.1 | - | 4 | 481 | 4792211 | ||
BC033026.1 | - | 4 | 416 |