Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144920.2 | NP_001138392.1 | 2 | 215 | ||
NM_001144923.2 | NP_001138395.1 | 2 | 215 | |||
NM_001287512.1 | NP_001274441.1 | 2 | 215 | |||
NM_001318333.1 | NP_001305262.1 | 2 | 215 | |||
NM_001321740.1 | NP_001308669.1 | 2 | 215 | |||
NM_001321741.1 | NP_001308670.1 | 2 | 215 | |||
NM_001321742.1 | NP_001308671.1 | 2 | 215 | |||
NM_024926.3 | NP_079202.2 | 2 | 215 | |||
GenBank mRNA | AK022633.1 | - | 2 | 181 | ||
AK295092.1 | - | 2 | 195 | |||
AK299132.1 | - | 2 | 176 | |||
AK300807.1 | - | 2 | 181 | |||
AL137393.1 | - | 2 | 179 | |||
BC013912.1 | - | 2 | 184 | |||
BC126331.1 | - | 2 | 140 | 8992047 | ||
BC130339.1 | - | 2 | 140 | 40146370 | ||
BP315162.1 | - | 2 | 215 |