Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK022953.1 | - | 1 | 699 | ||
AK092387.1 | - | 2 | 2637 | |||
AK096417.1 | - | 1 | 2016 |