Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPEF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024867.3 | NP_079143.3 | 28 | 4118 | ||
XM_005248376.3 | XP_005248433.1 | 28 | 4223 | |||
XM_005248377.3 | XP_005248434.1 | 28 | 4208 | |||
XM_011514135.2 | XP_011512437.1 | 28 | 4228 | |||
XM_011514136.2 | XP_011512438.1 | 28 | 4227 | |||
XM_011514137.2 | XP_011512439.1 | 28 | 4212 | |||
XM_011514138.2 | XP_011512440.1 | 28 | 4204 | |||
XM_011514139.2 | XP_011512441.1 | 27 | 4152 | |||
XM_011514140.2 | XP_011512442.1 | 27 | 3962 | |||
XM_011514141.2 | XP_011512443.1 | 29 | 4426 | |||
XM_011514142.1 | XP_011512444.1 | 28 | 4199 | |||
XM_017009880.1 | XP_016865369.1 | 27 | 4044 | |||
XM_017009881.1 | XP_016865370.1 | 28 | 5082 | |||
GenBank mRNA | AB051557.1 | - | 18 | 2459 | ||
AK027230.1 | - | 5 | 564 | |||
AK225586.1 | - | 5 | 564 | |||
AK225607.1 | - | 5 | 564 |