Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261394.1 | NP_001248323.1 | 4 | 406 | ||
NM_024894.3 | NP_079170.2 | 4 | 406 | |||
NR_048552.1 | 3 | 360 | ||||
XM_011510398.2 | XP_011508700.1 | 4 | 410 | |||
XM_011510399.2 | XP_011508701.1 | 3 | 348 | |||
XM_011510400.2 | XP_011508702.1 | 4 | 394 | |||
XM_011510402.2 | XP_011508704.1 | 4 | 394 | |||
XM_017004990.1 | XP_016860479.1 | 2 | 148 | |||
GenBank mRNA | AK024000.1 | - | 4 | 384 | ||
AK024137.1 | - | 4 | 391 | |||
AK290680.1 | - | 4 | 370 | |||
AK299617.1 | - | 3 | 348 | |||
BC005125.2 | - | 4 | 386 | 3841643 | ||
DC346574.1 | - | 4 | 406 |