Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144920.2 | NP_001138392.1 | 3 | 293 | ||
NM_001287512.1 | NP_001274441.1 | 3 | 293 | |||
NM_001287513.1 | NP_001274442.1 | 3 | 222 | |||
NM_001318333.1 | NP_001305262.1 | 3 | 293 | |||
NM_001321740.1 | NP_001308669.1 | 3 | 293 | |||
NM_001321741.1 | NP_001308670.1 | 3 | 293 | |||
NM_001321742.1 | NP_001308671.1 | 3 | 293 | |||
NM_024926.3 | NP_079202.2 | 3 | 293 | |||
GenBank mRNA | AK022633.1 | - | 3 | 259 | ||
AK293444.1 | - | 3 | 144 | |||
AK295092.1 | - | 3 | 273 | |||
AK300807.1 | - | 3 | 259 | |||
AL137393.1 | - | 3 | 257 | |||
BC013912.1 | - | 3 | 262 | |||
BC126331.1 | - | 3 | 218 | 8992047 | ||
BC130339.1 | - | 3 | 218 | 40146370 | ||
BP315162.1 | - | 3 | 293 |