Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM188A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318330.1 | NP_001305259.1 | 8, 9 | 991 | ||
NM_024948.3 | NP_079224.1 | 9, 10 | 1043 | |||
XM_005252600.2 | XP_005252657.1 | 8, 9 | 937 | |||
XM_005252602.2 | XP_005252659.1 | 7, 8 | 885 | |||
XM_011519690.2 | XP_011517992.1 | 9, 10 | 1020 | |||
XM_011519691.2 | XP_011517993.1 | 9, 10 | 1020 | |||
XM_011519693.1 | XP_011517995.1 | 9, 10 | 1029 | |||
XM_011519694.1 | XP_011517996.1 | 10, 11 | 1084 | |||
XM_017016672.1 | XP_016872161.1 | 11, 12 | 1176 | |||
XM_017016673.1 | XP_016872162.1 | 9, 10 | 1046 | |||
GenBank mRNA | AB014761.1 | - | 9, 10 | 809 | ||
AF063600.1 | - | 4, 5 | 309 | |||
AF176916.1 | - | 8, 9 | 953 | |||
AK023459.1 | - | 9, 10 | 1030 | |||
AL833873.1 | - | 9, 10 | 1005 | |||
BC020605.1 | - | 8, 9 | 956 | |||
BC067799.1 | - | 9, 10 | 1029 | 5278533 | ||
CR457315.1 | - | 9, 10 | 809 |