Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF556 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300843.1 | NP_001287772.1 | 3 | 233 | ||
NM_024967.2 | NP_079243.1 | 3 | 233 | |||
GenBank mRNA | AK021699.1 | - | 3 | 233 | ||
AK225297.1 | - | 3 | 233 | |||
BC009374.2 | - | 3 | 213 | 4121795 |