Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASRGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083926.1 | NP_001077395.1 | 2, 3 | 559 | ||
NM_025080.3 | NP_079356.3 | 2, 3 | 482 | |||
XM_011545265.2 | XP_011543567.1 | 2, 3 | 441 | |||
XM_011545266.2 | XP_011543568.1 | 2, 3 | 441 | |||
XM_017018354.1 | XP_016873843.1 | 2, 3 | 441 | |||
GenBank mRNA | AF411076.1 | - | 2, 3 | 291 | ||
AK090908.1 | - | 2, 3 | 331 | |||
AK091894.1 | - | 2, 3 | 413 | |||
AK225802.1 | - | 2, 3 | 250 | |||
AK313069.1 | - | 2, 3 | 342 | |||
BC021295.2 | - | 2, 3 | 280 | 3937243 | ||
BC093070.1 | - | 2, 3 | 384 | 30370741 | ||
BP308677.1 | - | 2, 3 | 482 |