Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASMTL-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026710.1 | 4 | 1099 | |||
GenBank mRNA | AF193043.2 | - | 3 | 462 | ||
BC093994.2 | - | 3 | 442 | |||
BC143527.1 | - | 3 | 476 |