Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319305.1 | NP_001306234.1 | 10 | 1048 | ||
NM_001319306.1 | NP_001306235.1 | 9 | 1003 | |||
NM_001319307.1 | NP_001306236.1 | 10 | 1066 | |||
XM_005263238.4 | XP_005263295.1 | 10 | 1099 | |||
XM_006714318.3 | XP_006714381.1 | 11 | 1144 | |||
XM_006714320.3 | XP_006714383.1 | 11 | 932 | |||
XM_006714322.3 | XP_006714385.1 | 7 | 482 | |||
XM_011532272.2 | XP_011530574.1 | 12 | 1211 | |||
XM_011532273.2 | XP_011530575.1 | 11 | 1166 | |||
XM_011532279.2 | XP_011530581.1 | 4 | 418 | |||
XM_017008632.1 | XP_016864121.1 | 4 | 332 | |||
GenBank mRNA | AK024759.1 | - | 9 | 815 | ||
BC111400.1 | - | 9 | 983 | |||
BC128142.1 | - | 9 | 753 | 40112521 |