Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATF7IP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256160.1 | NP_001243089.1 | 11, 12 | 1846 | ||
NM_024997.3 | NP_079273.2 | 11, 12 | 1870 | |||
NR_045815.1 | 9, 10 | 1644 | ||||
NR_045816.1 | 9, 10 | 1638 | ||||
XM_006720953.3 | XP_006721016.1 | 10, 11 | 1697 | |||
XM_011522666.2 | XP_011520968.1 | 7, 8 | 633 | |||
XM_017023705.1 | XP_016879194.1 | 10, 11 | 2838 | |||
XM_017023706.1 | XP_016879195.1 | 9, 10 | 2771 | |||
XM_017023708.1 | XP_016879197.1 | 9, 10 | 921 | |||
XM_017023709.1 | XP_016879198.1 | 6, 7 | 1232 | |||
XM_017023710.1 | XP_016879199.1 | 10, 11 | 1522 | |||
XM_017023711.1 | XP_016879200.1 | 6, 7 | 437 | |||
GenBank mRNA | AK022730.1 | - | 7, 8 | 630 | ||
AK093414.1 | - | 11, 12 | 1844 | |||
AK223492.1 | - | 7, 8 | 681 | |||
AK296565.1 | - | 5, 6 | 356 | |||
AK299040.1 | - | 9, 10 | 1629 | |||
AL832987.1 | - | 8, 9 | 1434 | |||
AY560615.1 | - | 9, 10 | 1640 | |||
BC033891.1 | - | 11, 12 | 1866 | 5266089 | ||
BC069695.1 | - | 7, 8 | 607 | |||
BC069713.1 | - | 7, 8 | 607 | |||
BC069730.1 | - | 7, 8 | 607 | |||
BC137079.1 | - | 10, 11 | 1769 | 9021076 | ||
BC144468.1 | - | 9, 10 | 1611 |