Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166133.1 | NP_001159605.1 | 37, 38 | 5417 | ||
NM_025074.6 | NP_079350.5 | 37, 38 | 5417 | |||
XM_006714316.2 | XP_006714379.1 | 37, 38 | 5664 | |||
GenBank mRNA | AK025684.1 | - | 4, 5 | 325 | ||
BC131820.1 | - | 37, 38 | 5007 |