Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASRGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083926.1 | NP_001077395.1 | 3 | 591 | ||
NM_025080.3 | NP_079356.3 | 3 | 514 | |||
XM_011545265.2 | XP_011543567.1 | 3 | 473 | |||
XM_011545266.2 | XP_011543568.1 | 3 | 473 | |||
XM_017018354.1 | XP_016873843.1 | 3 | 473 | |||
GenBank mRNA | AF411076.1 | - | 3 | 323 | ||
AK090908.1 | - | 3 | 363 | |||
AK091894.1 | - | 3 | 445 | |||
AK225802.1 | - | 3 | 282 | |||
AK313069.1 | - | 3 | 374 | |||
BC021295.2 | - | 3 | 312 | 3937243 | ||
BC093070.1 | - | 3 | 416 | 30370741 | ||
BP308677.1 | - | 3 | 514 |