Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CHD9 SIRNA Assays ›
Gene Symbol:
CHD9
Gene Name:
chromodomain helicase DNA binding protein 9
Gene Aliases:
AD013, CHD-9, CReMM, KISH2, PRIC320
Chromosome Location:
Chr.16: 53054991 - 53327502 on Build GRCh38
UniGene ID:
Hs.59159
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
32379
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CHD9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308319.1
NP_001295248.1
11
2897
NM_025134.4
NP_079410.4
12
2942
XM_005256168.3
XP_005256225.1
12
2984
XM_005256169.2
XP_005256226.1
12
3006
XM_005256172.3
XP_005256229.1
12
2923
XM_005256176.3
XP_005256233.1
11
1591
XM_006721281.2
XP_006721344.1
12
2923
XM_006721283.3
XP_006721346.1
12
3144
XM_011523347.2
XP_011521649.1
13
3056
XM_017023718.1
XP_016879207.1
13
3179
XM_017023719.1
XP_016879208.1
14
3130
XM_017023720.1
XP_016879209.1
13
3024
XM_017023721.1
XP_016879210.1
13
3180
XM_017023722.1
XP_016879211.1
13
3180
XM_017023723.1
XP_016879212.1
13
3180
XM_017023724.1
XP_016879213.1
13
3006
XM_017023725.1
XP_016879214.1
13
3006
XM_017023726.1
XP_016879215.1
11
2213
XM_017023727.1
XP_016879216.1
11
1591
XM_017023728.1
XP_016879217.1
13
3040
GenBank mRNA
AB002306.3
-
11
2365
AK022240.1
-
2
270
AK022582.1
-
1
80
AK299045.1
-
11
1640
AY243500.1
-
11
2820
AY647157.1
-
12
2942
BC140815.1
-
11
2774
9021683
BC144622.1
-
11
2774
9053153
DQ059482.1
-
2
270
DQ333316.1
-
12
3036
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_177224, AK086118, DQ127229, AK040994, AK030430, and others.
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
covalent chromatin modification
cellular lipid metabolic process
DNA binding
helicase activity
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0