Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025179.3 | NP_079455.3 | 31 | 6247 | ||
XM_005273164.2 | XP_005273221.1 | 32 | 6506 | |||
GenBank mRNA | AB007932.1 | - | 31 | 5698 | ||
AK021813.1 | - | 2 | 430 | |||
AK094367.1 | - | 2 | 1525 | |||
BC006193.2 | - | 9 | 1162 | |||
BC009343.2 | - | 14 | 2237 | |||
BC032125.2 | - | 2 | 364 | |||
BC132676.1 | - | 31 | 5602 | 40146698 | ||
BC136530.1 | - | 31 | 5602 | 9020520 |