Search Thermo Fisher Scientific
Gene Symbol: UBXN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171091.1 | NP_001164562.1 | 2, 3 | 249 | ||
NM_025241.2 | NP_079517.1 | 2, 3 | 351 | |||
XM_017027325.1 | XP_016882814.1 | 3, 4 | 782 | |||
GenBank mRNA | AF272893.1 | - | 2, 3 | 350 | ||
AF272894.1 | - | 2, 3 | 240 | |||
BC008288.1 | - | 2, 3 | 249 | 3659797 | ||
BM764980.1 | - | 2, 3 | 351 |