Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195059.1 | NP_001181988.1 | 5 | 2916 | ||
NM_025201.4 | NP_079477.2 | 6 | 3066 | |||
GenBank mRNA | AF217986.1 | - | 1 | 1214 | ||
AF318373.1 | - | 3 | 2664 | |||
AF494534.1 | - | 6 | 3015 | |||
AK096203.1 | - | 7 | 1737 | |||
AL157457.1 | - | 1 | 796 | |||
BC008744.2 | - | 5 | 2877 | 3346334 |