Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGES3L-AARSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136042.2 | NP_001129514.2 | 17 | 2147 | ||
NM_025267.3 | NP_079543.1 | 17 | 1763 | |||
GenBank mRNA | AF039235.1 | - | 17 | 1851 | ||
AK094084.1 | - | 17 | 1701 | |||
BC004172.2 | - | 17 | 1763 | 2823004 | ||
BC019324.1 | - | 17 | 1763 | 2823004 |
Gene Symbol: AARSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261434.1 | NP_001248363.1 | 12 | 1337 |