Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030613.3 | NP_085116.2 | 5 | 1657 | ||
GenBank mRNA | AK025281.1 | - | 1 | 972 | ||
AK131367.1 | - | 5 | 1636 | |||
BC132779.1 | - | 3 | 1430 | 40146801 | ||
BC132781.1 | - | 3 | 1455 | 40146803 | ||
BC142989.1 | - | 5 | 1626 | 8860436 | ||
BC144213.1 | - | 3 | 1430 | 9052733 | ||
BC144216.1 | - | 4 | 1541 | 9052736 |