Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045104.1 | 2 | 331 | |||
NR_045105.2 | 2 | 585 | ||||
NR_045106.2 | 2 | 492 | ||||
NR_045901.1 | 2 | 331 | ||||
XM_006720708.3 | XP_006720771.1 | 2 | 267 | |||
GenBank mRNA | AK027475.1 | - | 2 | 173 | ||
AK296134.1 | - | 2 | 558 | |||
AK301222.1 | - | 2 | 284 | |||
AL136662.1 | - | 2 | 413 | |||
BC007991.2 | - | 2 | 158 | 3140504 | ||
BC037823.1 | - | 1 | 883 | |||
BP315446.1 | - | 2 | 480 | |||
DC313962.1 | - | 2 | 476 |