Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030775.2 | NP_110402.2 | 3 | 351 | ||
NM_032642.2 | NP_116031.1 | 3 | 389 | |||
GenBank mRNA | AB060966.1 | - | 2 | 211 | ||
AB209228.1 | - | 2 | 749 | |||
AK290430.1 | - | 3 | 450 | |||
AY009399.1 | - | 3 | 351 | |||
BC001749.1 | - | 3 | 389 | 3528819 |