Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030793.4 | NP_110420.3 | 15 | 2533 | ||
XM_005268513.1 | XP_005268570.1 | 15 | 2522 | |||
XM_006714797.1 | XP_006714860.1 | 15 | 2522 | |||
XM_011537684.2 | XP_011535986.1 | 15 | 2575 | |||
XM_017009899.1 | XP_016865388.1 | 16 | 2670 | |||
XM_017009900.1 | XP_016865389.1 | 16 | 2734 | |||
XM_017009901.1 | XP_016865390.1 | 15 | 2587 | |||
XM_017009902.1 | XP_016865391.1 | 15 | 2575 | |||
GenBank mRNA | AF177339.1 | - | 1 | 454 | ||
AF251055.1 | - | 8 | 1471 | |||
AK313558.1 | - | 2 | 612 | |||
AL832841.1 | - | 5 | 1000 | |||
BC005849.2 | - | 5 | 1107 | |||
BC005873.2 | - | 5 | 1107 | |||
BC015366.1 | - | 1 | 179 | |||
BC033454.1 | - | 15 | 2466 | 4510274 | ||
BC056147.1 | - | 1 | 359 | |||
BC078159.1 | - | 1 | 199 |