Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM49A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030797.3 | NP_110424.1 | 4 | 390 | ||
XM_005262630.3 | XP_005262687.1 | 3 | 346 | |||
XM_006712108.3 | XP_006712171.1 | 4 | 345 | |||
GenBank mRNA | AK055334.1 | - | 4 | 358 | ||
AK315065.1 | - | 3 | 285 | |||
AL136704.1 | - | 4 | 355 | |||
BC038971.1 | - | 4 | 341 | 5200807 | ||
CR533504.1 | - | 2 | 137 | |||
DA188616.1 | - | 4 | 390 |