Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045104.1 | 2, 3 | 484 | |||
NR_045105.2 | 2, 3 | 738 | ||||
NR_045106.2 | 2, 3 | 645 | ||||
XM_006720708.3 | XP_006720771.1 | 2, 3 | 420 | |||
GenBank mRNA | AK027475.1 | - | 2, 3 | 326 | ||
AK296134.1 | - | 2, 3 | 711 | |||
AK310857.1 | - | 2, 3 | 330 | |||
AL136662.1 | - | 2, 3 | 566 | |||
BC007991.2 | - | 2, 3 | 311 | 3140504 |