Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008695.1 | NP_001008695.1 | 5 | 920 | ||
NM_030573.2 | NP_085050.2 | 4 | 1094 | |||
GenBank mRNA | AK024609.1 | - | 2 | 1661 | ||
AK025906.1 | - | 2 | 1426 | |||
AK300715.1 | - | 5 | 709 | |||
AK315457.1 | - | 4 | 1079 | |||
BC004346.1 | - | 4 | 1065 | 3633743 | ||
CR456343.1 | - | 4 | 912 |