Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024938.3 | NP_001020109.1 | 12 | 1683 | ||
NM_001024939.3 | NP_001020110.1 | 12 | 1571 | |||
NM_001282864.1 | NP_001269793.1 | 11 | 1925 | |||
NM_030807.4 | NP_110434.3 | 13 | 1794 | |||
NR_104247.1 | 11 | 1530 | ||||
GenBank mRNA | AB209301.1 | - | 8 | 2156 | ||
AF443201.1 | - | 13 | 1625 | |||
AJ271290.1 | - | 12 | 1514 | |||
AK075467.1 | - | 12 | 1459 | |||
AK092466.1 | - | 2 | 1595 | |||
AK314502.1 | - | 13 | 1565 | |||
BC018967.2 | - | 1 | 239 | |||
BC094735.1 | - | 12 | 1500 | 5304025 | ||
BC100807.2 | - | 9 | 1632 | |||
BC100808.2 | - | 12 | 1429 | 40000757 | ||
BC100809.1 | - | 12 | 1429 | 40000760 | ||
BC100810.1 | - | 11 | 1276 | |||
CR456373.1 | - | 12 | 1460 |