Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD13C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030816.4 | NP_110443.3 | 9 | 1449 | ||
XM_005271234.3 | XP_005271291.1 | 9 | 1692 | |||
XM_005271235.3 | XP_005271292.1 | 9 | 1439 | |||
XM_006710929.3 | XP_006710992.1 | 9 | 1462 | |||
XM_017002413.1 | XP_016857902.1 | 7 | 672 | |||
XM_017002414.1 | XP_016857903.1 | 8 | 754 | |||
XM_017002415.1 | XP_016857904.1 | 8 | 733 | |||
XM_017002416.1 | XP_016857905.1 | 4 | 418 | |||
GenBank mRNA | AK074187.1 | - | 1 | 73 | ||
AL136717.1 | - | 8 | 1345 | |||
BC028840.1 | - | 9 | 1438 | 4558748 |