Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RNF32 SIRNA Assays ›
Gene Symbol:
RNF32
Gene Name:
ring finger protein 32
Gene Aliases:
FKSG33, HSD15, LMBR2
Chromosome Location:
Chr.7: 156640286 - 156678333 on Build GRCh38
UniGene ID:
Hs.446194
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
32933
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RNF32
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001184996.1
NP_001171925.1
3
514
NM_001184997.1
NP_001171926.1
3
367
NM_001308273.1
NP_001295202.1
3
465
NM_001308274.1
NP_001295203.1
2
256
NM_030936.3
NP_112198.1
3
409
NR_131778.1
2
256
XM_005249522.4
XP_005249579.1
3
321
XM_011515804.2
XP_011514106.1
4
431
XM_011515805.2
XP_011514107.1
4
588
XM_011515806.2
XP_011514108.1
3
499
XM_011515807.2
XP_011514109.1
3
307
XM_011515808.2
XP_011514110.1
3
419
XM_011515809.2
XP_011514111.1
3
492
XM_011515810.2
XP_011514112.1
3
309
XM_011515811.2
XP_011514113.1
3
304
XM_011515812.2
XP_011514114.1
3
411
XM_011515813.2
XP_011514115.1
3
409
XM_017011753.1
XP_016867242.1
3
413
XM_017011754.1
XP_016867243.1
3
413
XM_017011755.1
XP_016867244.1
3
413
GenBank mRNA
AA412402.1
-
3
377
AF325690.1
-
3
317
AF441222.1
-
2
256
AF441224.1
-
2
256
AK307316.1
-
3
374
AK314067.1
-
3
514
AL136874.1
-
3
297
BC015416.1
-
2
267
4395421
BI460257.1
-
3
412
BI465216.1
-
3
345
BT007037.1
-
1
179
CR533513.1
-
1
179
DB198291.1
-
3
514
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0