Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031280.3 | NP_112570.2 | 3, 4 | 424 | ||
GenBank mRNA | AB049946.1 | - | 3, 4 | 299 | ||
AB209905.1 | - | 3, 4 | 323 | |||
AF265439.1 | - | 3, 4 | 343 | |||
AK302633.1 | - | 3, 4 | 368 | |||
AK315441.1 | - | 3, 4 | 363 | |||
BC031336.1 | - | 3, 4 | 339 | 3543896 | ||
BF310592.1 | - | 3, 4 | 357 | |||
BM048518.1 | - | 3, 4 | 423 | |||
BU927793.1 | - | 3, 4 | 274 |