Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAB39L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079670.2 | NP_001073138.1 | 7, 8 | 958 | ||
NM_001287337.1 | NP_001274266.1 | 8, 9 | 1096 | |||
NM_001287338.1 | NP_001274267.1 | 8, 9 | 1091 | |||
NM_001287339.1 | NP_001274268.1 | 6, 7 | 760 | |||
NM_030925.3 | NP_112187.2 | 5, 6 | 1009 | |||
XM_011535254.2 | XP_011533556.1 | 9, 10 | 1500 | |||
XM_011535255.2 | XP_011533557.1 | 7, 8 | 980 | |||
XM_011535256.2 | XP_011533558.1 | 8, 9 | 1245 | |||
XM_017020785.1 | XP_016876274.1 | 8, 9 | 1372 | |||
XM_017020786.1 | XP_016876275.1 | 9, 10 | 1509 | |||
XM_017020787.1 | XP_016876276.1 | 9, 10 | 1762 | |||
XM_017020788.1 | XP_016876277.1 | 10, 11 | 1638 | |||
GenBank mRNA | AA278473.1 | - | 3, 4 | 445 | ||
AK022639.1 | - | 4, 5 | 554 | |||
AK309011.1 | - | 2, 3 | 284 | |||
AY288977.2 | - | 8, 9 | 1023 | |||
BC010993.2 | - | 5, 6 | 834 | 3956127 | ||
BX647518.1 | - | 7, 8 | 895 |