Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030939.4 | NP_112201.1 | 2 | 677 | ||
XM_005249433.3 | XP_005249490.1 | 2 | 1084 | |||
XM_011514928.2 | XP_011513230.1 | 2 | 1018 | |||
GenBank mRNA | AB075502.1 | - | 2 | 664 | ||
AK022681.1 | - | 2 | 561 | |||
AK301718.1 | - | 2 | 216 | |||
AL136632.2 | - | 2 | 688 | |||
AY598792.1 | - | 2 | 170 | |||
BC040260.1 | - | 2 | 189 | |||
BC047866.1 | - | 2 | 368 | 6499979 | ||
CR457292.1 | - | 2 | 170 | |||
CR533486.1 | - | 2 | 170 |