Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITM2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012514.2 | NP_001012532.1 | 5 | 1241 | ||
NM_001012516.2 | NP_001012534.1 | 5 | 1216 | |||
NM_001287240.1 | NP_001274169.1 | 6 | 1318 | |||
NM_001287241.1 | NP_001274170.1 | 7 | 1514 | |||
NM_030926.5 | NP_112188.1 | 6 | 1382 | |||
GenBank mRNA | AF271781.1 | - | 6 | 1367 | ||
AF272043.1 | - | 6 | 1364 | |||
AK056321.1 | - | 5 | 1188 | |||
AK075361.1 | - | 6 | 1347 | |||
AK090975.1 | - | 6 | 1293 | |||
AK126739.1 | - | 1 | 2462 | |||
AK222697.1 | - | 6 | 1329 | |||
AK307702.1 | - | 5 | 1150 | |||
AL136603.1 | - | 6 | 1323 | |||
AL713651.1 | - | 6 | 1306 | |||
BC002424.1 | - | 6 | 1309 | 3162430 | ||
BC025742.1 | - | 5 | 1185 | 5224043 | ||
BC050668.1 | - | 5 | 1176 | 5759920 | ||
BC098563.1 | - | 6 | 1295 | 30558635 | ||
CR749389.1 | - | 1 | 2420 | |||
D87062.1 | - | 1 | 329 |