Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184996.1 | NP_001171925.1 | 9 | 1445 | ||
NM_001184997.1 | NP_001171926.1 | 9 | 1298 | |||
NM_001308273.1 | NP_001295202.1 | 9 | 1520 | |||
NM_030936.3 | NP_112198.1 | 9 | 1340 | |||
NR_131778.1 | 9 | 1294 | ||||
XM_005249522.4 | XP_005249579.1 | 9 | 1252 | |||
XM_011515804.2 | XP_011514106.1 | 10 | 1362 | |||
XM_011515805.2 | XP_011514107.1 | 10 | 1519 | |||
XM_011515806.2 | XP_011514108.1 | 9 | 1430 | |||
XM_011515807.2 | XP_011514109.1 | 9 | 1238 | |||
XM_011515808.2 | XP_011514110.1 | 9 | 1350 | |||
XM_011515809.2 | XP_011514111.1 | 9 | 1423 | |||
XM_011515810.2 | XP_011514112.1 | 9 | 1240 | |||
XM_011515811.2 | XP_011514113.1 | 9 | 1235 | |||
XM_011515812.2 | XP_011514114.1 | 9 | 1342 | |||
XM_011515813.2 | XP_011514115.1 | 9 | 1301 | |||
GenBank mRNA | AF325690.1 | - | 9 | 1248 | ||
AF441222.1 | - | 8 | 1187 | |||
AF441224.1 | - | 9 | 1294 | |||
AL136874.1 | - | 9 | 1228 | |||
CR533513.1 | - | 7 | 1110 |