Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSDMC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031415.2 | NP_113603.1 | 2 | 1019 | ||
XM_011517158.1 | XP_011515460.1 | 2 | 759 | |||
XM_011517159.1 | XP_011515461.1 | 2 | 759 | |||
XM_011517161.1 | XP_011515463.1 | 2 | 759 | |||
XM_011517162.1 | XP_011515464.1 | 2 | 759 | |||
XM_011517167.1 | XP_011515469.1 | 2 | 759 | |||
XM_011517168.1 | XP_011515470.1 | 2 | 759 | |||
XM_017013644.1 | XP_016869133.1 | 2 | 759 | |||
GenBank mRNA | AB042405.1 | - | 2 | 1028 | ||
BC035321.1 | - | 2 | 564 | 4753282 | ||
BC063595.1 | - | 2 | 547 | 4752106 |