Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005392.3 | NP_005383.3 | 22 | 4671 | ||
XM_005252051.2 | XP_005252108.1 | 22 | 4668 | |||
XM_006717143.2 | XP_006717206.1 | 22 | 4563 | |||
GenBank mRNA | AB014562.2 | - | 21 | 4579 | ||
AL834263.1 | - | 22 | 4538 | |||
BC050112.1 | - | 3 | 1903 | |||
BC064613.1 | - | 9 | 2259 | |||
BC152437.1 | - | 21 | 4579 |