Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313726.1 | NP_001300655.1 | 3 | 440 | ||
NM_031418.3 | NP_113606.2 | 2 | 348 | |||
GenBank mRNA | AJ300461.1 | - | 2 | 1055 | ||
AK302659.1 | - | 2 | 242 | |||
AK304781.1 | - | 3 | 281 | |||
DA247850.1 | - | 2 | 308 |