Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031421.3 | NP_113609.1 | 3 | 370 | ||
NR_110662.1 | 3 | 370 | ||||
XM_011525324.2 | XP_011523626.1 | 3 | 337 | |||
GenBank mRNA | AK055498.1 | - | 3 | 315 | ||
AL136760.1 | - | 3 | 337 | |||
BC025390.1 | - | 3 | 378 | |||
BC067297.1 | - | 3 | 329 | 30383608 |