Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031439.3 | NP_113627.1 | 2 | 1796 | ||
GenBank mRNA | AK055556.1 | - | 2 | 1734 | ||
BC004299.1 | - | 2 | 1723 | |||
BC071947.1 | - | 2 | 1722 | 6459570 | ||
BU187815.1 | - | 1 | 115 |