Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19orf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031726.3 | NP_001026896.2 | 3 | 1311 | ||
NM_001256046.1 | NP_001242975.1 | 4 | 1527 | |||
NM_001256047.1 | NP_001242976.1 | 3 | 1388 | |||
NM_001282929.1 | NP_001269858.1 | 2 | 1141 | |||
NM_001282930.1 | NP_001269859.1 | 2 | 1412 | |||
NM_001282931.1 | NP_001269860.1 | 4 | 1703 | |||
NM_031448.4 | NP_113636.2 | 3 | 1582 | |||
GenBank mRNA | AK057185.1 | - | 3 | 1279 | ||
AL162066.1 | - | 2 | 1086 | |||
BC004957.1 | - | 2 | 1072 | 3628073 | ||
BC009946.2 | - | 2 | 1072 | 4125477 | ||
BC017211.2 | - | 1 | 972 | |||
BC063518.1 | - | 4 | 1206 | 4340755 |