Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282658.1 | NP_001269587.1 | 6 | 1162 | ||
NM_031455.3 | NP_113643.1 | 2 | 597 | |||
GenBank mRNA | AK095792.1 | - | 6 | 1041 | ||
AL136562.1 | - | 2 | 513 | |||
BC051334.1 | - | 2 | 597 | 6302570 |