Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077442.1 | NP_001070910.1 | 8 | 1525 | ||
NM_001077443.1 | NP_001070911.1 | 8 | 1486 | |||
NM_004500.3 | NP_004491.2 | 9 | 1512 | |||
NM_031314.2 | NP_112604.2 | 9 | 1551 | |||
XM_006720125.2 | XP_006720188.1 | 9 | 2168 | |||
XM_011536708.1 | XP_011535010.1 | 10 | 1585 | |||
XM_011536709.2 | XP_011535011.1 | 9 | 2006 | |||
XM_011536710.2 | XP_011535012.1 | 9 | 2207 | |||
XM_011536711.2 | XP_011535013.1 | 9 | 2214 | |||
XM_011536712.1 | XP_011535014.1 | 10 | 1546 | |||
XM_017021252.1 | XP_016876741.1 | 9 | 2175 | |||
XM_017021253.1 | XP_016876742.1 | 9 | 1966 | |||
GenBank mRNA | AK126950.1 | - | 8 | 4220 | ||
AK223517.1 | - | 9 | 1490 | |||
AK302213.1 | - | 7 | 1548 | |||
AL833432.1 | - | 6 | 1089 | |||
BC003394.1 | - | 9 | 1443 | 3447724 | ||
BC066932.1 | - | 8 | 1431 | 4375450 | ||
BC092433.1 | - | 5 | 887 | |||
BC103758.1 | - | 8 | 1431 | 6187512 | ||
BX247961.1 | - | 8 | 1242 | |||
BX247992.1 | - | 8 | 1397 | |||
M16342.1 | - | 9 | 1388 |