Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMMECR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199140.1 | NP_001186069.1 | 8 | 4323 | ||
NM_031445.2 | NP_113633.2 | 8 | 4295 | |||
XM_005263804.4 | XP_005263861.1 | 9 | 4926 | |||
XM_005263806.4 | XP_005263863.1 | 8 | 4815 | |||
XM_005263807.4 | XP_005263864.1 | 7 | 4680 | |||
XM_011511952.2 | XP_011510254.1 | 8 | 4816 | |||
XM_011511954.2 | XP_011510256.1 | 9 | 4903 | |||
XM_011511955.2 | XP_011510257.1 | 7 | 3845 | |||
GenBank mRNA | AK024993.1 | - | 1 | 1770 | ||
AK026685.1 | - | 1 | 2656 | |||
AL137415.1 | - | 1 | 454 | |||
BC016181.1 | - | 8 | 4295 | 4644159 | ||
BC078174.1 | - | 8 | 4305 | 6012672 | ||
BC084552.1 | - | 1 | 643 |