Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SH3KBP1 SIRNA Assays ›
Gene Symbol:
SH3KBP1
Gene Name:
SH3 domain containing kinase binding protein 1
Gene Aliases:
CD2BP3, CIN85, GIG10, HSB-1, HSB1, MIG18
Chromosome Location:
Chr.X: 19533965 - 19887626 on Build GRCh38
UniGene ID:
Hs.726365
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
33440
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SH3KBP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001024666.2
NP_001019837.1
7
883
NM_001184960.1
NP_001171889.1
3
293
NM_031892.2
NP_114098.1
8
1195
XM_005274494.2
XP_005274551.1
9
1136
XM_011545498.2
XP_011543800.1
10
1193
XM_011545499.2
XP_011543801.1
9
1001
XM_011545500.2
XP_011543802.1
10
1193
XM_011545502.2
XP_011543804.1
3
299
XM_011545503.2
XP_011543805.1
10
1193
XM_017029459.1
XP_016884948.1
9
933
XM_017029460.1
XP_016884949.1
9
976
XM_017029461.1
XP_016884950.1
8
919
XM_017029462.1
XP_016884951.1
9
1136
XM_017029463.1
XP_016884952.1
7
903
XM_017029464.1
XP_016884953.1
10
1268
XM_017029465.1
XP_016884954.1
8
1162
XM_017029466.1
XP_016884955.1
8
1061
XM_017029467.1
XP_016884956.1
9
1210
XM_017029468.1
XP_016884957.1
8
1162
XM_017029469.1
XP_016884958.1
3
249
XM_017029470.1
XP_016884959.1
3
401
GenBank mRNA
AF230904.1
-
8
1170
AF329267.1
-
2
96
AF329268.1
-
2
96
AF542051.1
-
7
846
AK293234.1
-
7
823
AK293237.1
-
3
193
AK300189.1
-
3
203
AK313308.1
-
8
1123
AY423734.1
-
9
1093
AY542305.1
-
8
930
BC015806.1
-
8
1097
3906722
BC050663.2
-
8
1105
BP224209.1
-
3
292
DC359739.1
-
3
296
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
apoptotic process
cytoskeleton organization
cell-cell signaling
regulation of cell shape
cell migration
negative regulation of epidermal growth factor receptor signaling pathway
protein binding
SH3 domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0