Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GORASP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278789.1 | NP_001265718.1 | 7 | 2852 | ||
NM_001278790.1 | NP_001265719.1 | 6 | 2672 | |||
NM_031899.3 | NP_114105.1 | 9 | 3600 | |||
NR_103866.1 | 9 | 3096 | ||||
NR_103867.1 | 9 | 3088 | ||||
XM_006713301.3 | XP_006713364.1 | 9 | 3700 | |||
XM_011534020.2 | XP_011532322.1 | 9 | 3610 | |||
XM_011534021.2 | XP_011532323.1 | 8 | 3520 | |||
XM_017007050.1 | XP_016862539.1 | 9 | 2786 | |||
XM_017007051.1 | XP_016862540.1 | 8 | 3135 | |||
XM_017007052.1 | XP_016862541.1 | 8 | 2731 | |||
GenBank mRNA | AJ409349.1 | - | 9 | 2876 | ||
AK001574.1 | - | 8 | 3118 | |||
AK027096.1 | - | 1140 | ||||
AK057043.1 | - | 6 | 2410 | |||
AK091168.1 | - | 10 | 2531 | |||
AK124755.1 | - | 9 | 3600 | |||
BC008928.2 | - | 9 | 2817 | |||
BC075854.1 | - | 9 | 2886 | 5503263 |