Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032120.3 | NP_115496.2 | 2 | 229 | ||
XM_005250636.4 | XP_005250693.1 | 2 | 293 | |||
XM_006716149.3 | XP_006716212.1 | 2 | 244 | |||
XM_006716150.3 | XP_006716213.1 | 2 | 244 | |||
XM_017012709.1 | XP_016868198.1 | 1 | 77 | |||
GenBank mRNA | AF161422.1 | - | 2 | 196 | ||
AK294626.1 | - | 2 | 203 | |||
AK294812.1 | - | 2 | 201 | |||
AL136619.1 | - | 2 | 211 | |||
BC020919.1 | - | 2 | 207 | |||
BC033561.1 | - | 2 | 208 | |||
BC044570.1 | - | 2 | 212 | 4793676 | ||
BC065716.1 | - | 2 | 199 | |||
BC104865.1 | - | 2 | 229 | 8143868 | ||
BC104867.1 | - | 2 | 229 | 8143870 | ||
BC143576.1 | - | 2 | 230 | 9052087 | ||
CR533549.1 | - | 2 | 188 | |||
CR933641.1 | - | 2 | 217 | |||
CR936726.1 | - | 2 | 208 |