Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000635.3 | NP_000626.2 | 10, 11 | 1411 | ||
NM_134433.2 | NP_602309.1 | 9, 10 | 1336 | |||
XM_011528171.2 | XP_011526473.1 | 11, 12 | 1377 | |||
XM_011528172.1 | XP_011526474.1 | 10, 11 | 1234 | |||
XM_017027107.1 | XP_016882596.1 | 9, 10 | 1261 | |||
XM_017027108.1 | XP_016882597.1 | 8, 9 | 1186 | |||
GenBank mRNA | AK024288.1 | - | 7, 8 | 950 | ||
AK291196.1 | - | 9, 10 | 1215 | |||
AK302661.1 | - | 4, 5 | 399 | |||
AK303321.1 | - | 7, 8 | 720 | |||
BC028579.1 | - | 10, 11 | 1323 | 4829582 | ||
BC071571.1 | - | 10, 11 | 1292 | 5276369 | ||
BX648130.1 | - | 10, 11 | 1246 | |||
X76091.1 | - | 10, 11 | 1301 |