Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETDB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF334407.1 | - | 7 | 1182 | ||
AK307593.1 | - | 5 | 576 | |||
AK308220.1 | - | 6 | 1146 | |||
BC047434.2 | - | 5 | 663 | 5266911 |