Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005230.3 | NP_005221.2 | 3 | 556 | ||
XM_006719275.3 | XP_006719338.1 | 4 | 653 | |||
GenBank mRNA | AK312694.1 | - | 3 | 467 | ||
BC017371.1 | - | 3 | 438 | 4287696 | ||
CR542251.1 | - | 2 | 230 | |||
Z36715.1 | - | 3 | 509 |